Ethyl-3-hydroxybutyrat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Ethyl 3-hydroxybutanoate, DL-Ethyl-3-hydroxybutyrate, Ethyl 3-hydroxybutyrate

M-Clarity™ Qualitätsstufe = MQ300

W342807-100G-K W342807-10KG-K W342807-1KG-K
SIALW342807-100G-KEA 89.3 EUR
SIALW342807-100G-K SIALW342807-10KG-K SIALW342807-1KG-K
Ethyl-3-hydroxybutyrat, Sigma-Aldrich®
Ethyl-3-hydroxybutyrat
Formel: C₆H₁₂O₃
Molecular Weight: 132,16 g/mol
MDL: MFCD00004545
CAS-Nummer: 5405-41-4
EINECS: 226-456-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR