Bis(2-chlorethyl)ammoniumchlorid 98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Bis(2-chloroethyl)amine hydrochloride
A13118.0B
A13118.18
A13118.30
A13118.0BEA
192
EUR
A13118.0B
A13118.18
A13118.30
Bis(2-chlorethyl)ammoniumchlorid 98%
Bis(2-chlorethyl)ammoniumchlorid
Purity: 98%
CAS No.: 821-48-7
Molecular Formula: C4H9Cl2N.HCl
Formula Weight: 178.49
Melting Point: 211-215°
Beilstein Reference: 3550356
UN No.: 2923
MDL No.: MFCD00012515
Formel:
C₄H₁₀Cl₃N Molecular Weight: 178,49 g/mol Schmelzpunkt: 211…215 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00012515 CAS-Nummer: 821-48-7 EINECS: 212-479-5 UN: 2923 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...