2,5-Dichlor-3-thenoesäure, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
2,5-Dichlorothiophene-3-carboxylic acid
M-Clarity™ Qualitätsstufe = MQ100
654795-25G
654795-5G
SIAL654795-25GEA
208
EUR
SIAL654795-25G
SIAL654795-5G
2,5-Dichlor-3-thenoesäure, Sigma-Aldrich®
2,5-Dichlor-3-thenoesäure
Formel:
C₅H₂Cl₂O₂S Molecular Weight: 197,04 g/mol |
MDL:
MFCD00082908 CAS-Nummer: 36157-41-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...