Glycin-tert-butylester (H-Gly-OtBu) ≥97%

Lieferant: Thermo Fisher Scientific

Synonyme: tert-Butyl 2-aminoacetate, tert-Butyl Glycinate

L16258.03 L16258.06
L16258.03EA 66.9 EUR
L16258.03 L16258.06
Glycin-tert-butylester (H-Gly-OtBu) ≥97%
Glycin-tert-butylester (H-Gly-OtBu)

Purity: 97% CAS No.: 6456-74-2 Molecular Formula: H2NCH2CO2C(CH3)3 Formula Weight: 131.18 Storage Temperature: 2°C to 8°C Refractive Index: 1.4240 Boiling Point: 29-31°/2mm Beilstein Reference: 2039000 MDL No.: MFCD00038194

Formel: C₆H₁₃NO₂
Siedepunkt: 29…31 °C (2 mmHg)
Dichte: 0,960
MDL: MFCD00038194
CAS-Nummer: 6456-74-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR