1-(4-Nitrophenyl)pyrrolidin

Lieferant: BIONET RESEARCH

10G-310S.500 10G-310S.1 10G-310S.5 10G-310S.10
BIOT10G-310S.500EA 0 EUR
BIOT10G-310S.500 BIOT10G-310S.1 BIOT10G-310S.5 BIOT10G-310S.10
1-(4-Nitrophenyl)pyrrolidin
1-(4-Nitrophenyl)pyrrolidin
Formel: C₁₀H₁₂N₂O₂
MDL: MFCD00020819
CAS-Nummer: 10220-22-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR