3-Hydroxy-3-methylbuttersäure ≥96%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
3-Hydroxyisovaleric acid, 3-Hydroxy-3-methylbutanoic acid
42722.18
42722.30
42722.09
42722.18EA
133
EUR
42722.18
42722.30
42722.09
3-Hydroxy-3-methylbuttersäure ≥96%
3-Hydroxy-3-methylbuttersäure
Purity: 96%
CAS No.: 625-08-1
Molecular Formula: C5H10O3
Formula Weight: 118.13
Flash Point: 113°(235°F)
Physical Form: Liquid
Density: 1.1
Refractive Index: 1.4408
Appearance: Pale yellow
Boiling Point: 128°/7mm
Solubility: Soluble in water
Beilstein Reference: 1743952
MDL No.: MFCD00059081
Formel:
C₅H₁₀O₃ Molecular Weight: 118.13 g/mol Siedepunkt: 128 °C (7 mmHg) Dichte: 1.1 Flammpunkt: 113 °C (235 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00059081 CAS-Nummer: 625-08-1 |