2-Chlor-6-fluorphenylacetonitril
Lieferant: FLUOROCHEM
005668-1G
005668-5G
FLUO005668-1GEA
31.1
EUR
FLUO005668-1G
FLUO005668-5G
2-Chlor-6-fluorphenylacetonitril
2-Chlor-6-fluorphenylacetonitril
Formel:
C₈H₅ClFN |
MDL:
MFCD00001900 CAS-Nummer: 75279-55-9 EINECS: 278-171-8 UN: 3439 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...