4-Chlor-2-fluorphenylacetonitril
Lieferant: FLUOROCHEM
Synonyme:
4-Chloro-2-fluorobenzeneacetonitrile
009421-1G
009421-5G
009421-25G
FLUO009421-1GEA
0
EUR
FLUO009421-1G
FLUO009421-5G
FLUO009421-25G
4-Chlor-2-fluorphenylacetonitril
4-Chlor-2-fluorphenylacetonitril
Formel:
C₈H₅ClFN Molecular Weight: 169.59 g/mol |
MDL:
MFCD01312758 CAS-Nummer: 75279-53-7 UN: 3439 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...