Kaliumtetrachloroaurat Hydrat ≥99,99% (Metall-Basis) Au ≥ 49%, Premion®
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
Gold potassium chloride hydrate, Potassium tetrachloroaurate(III) hydrate, Potassium gold(III) chloride hydrate
12150.03
12150.06
12150.03EA
149
EUR
12150.03
12150.06
Kaliumtetrachloroaurat Hydrat ≥99,99% (Metall-Basis) Au ≥ 49%, Premion®
Kaliumtetrachloroaurat Hydrat
Purity: 99.99%
CAS No.: 27988-75-6
Molecular Formula: AuCl4K.xH2O
Formula Weight: 377.88(anhy)
Physical Form: Crystalline
Appearance: Yellow to orange
Melting Point: 357° dec.
Solubility: Soluble in water
Merck Reference: 14,7681
UN No.: 3260
MDL No.: MFCD00043088
Formel:
AuCl₄K·xH₂O Molecular Weight: 377,88 g/mol (anhy) Schmelzpunkt: 357 °C (Zersetzung) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00043088 CAS-Nummer: 27988-75-6 EINECS: 237-190-1 UN: 3260 ADR: 8,III Merck Index: 14,7681 |