Weißes Mineralöl (Erdöl), M.W. 75 cSt

Lieferant: JOBIN YVON EMISSION

Synonyme: Matrix oil, Mineral oil, Paraffin oil

BASE75 BASE75-G
JOBIBASE75EA 146 EUR
JOBIBASE75 JOBIBASE75-G
Weißes Mineralöl (Erdöl), M.W. 75 cSt
Mineral oil, white (petroleum)
MDL: MFCD00131611
CAS-Nummer: 8042-47-5
EINECS: 232-455-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR