3-Fluor-4-methoxybenzylchlorid 98%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
3-Fluoro-4-methoxybenzylchloride
H31756.03
H31756.06
H31756.03EA
56.1
EUR
H31756.03
H31756.06
3-Fluor-4-methoxybenzylchlorid 98%
3-Fluor-4-methoxybenzylchlorid
Purity: 98%
CAS No.: 351-52-0
Molecular Formula: C8H8ClFO
Formula Weight: 174.60
Melting Point: 35-39°
UN No.: 3261
MDL No.: MFCD04973773
Formel:
C₈H₈ClFO Molecular Weight: 174,60 g/mol Schmelzpunkt: 35…39 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04973773 CAS-Nummer: 351-52-0 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...