3-Fluor-4-methoxybenzylchlorid

Lieferant: CHEMPUR

Synonyme: 3-Fluoro-4-methoxybenzylchloride

FL13840.POR 013840-5G 013840-25G
CHMPFL13840.POREA 0 EUR
CHMPFL13840.POR FLUO013840-5G FLUO013840-25G
3-Fluor-4-methoxybenzylchlorid
3-Fluor-4-methoxybenzylchlorid
Formel: C₈H₈ClFO
Molecular Weight: 174,60 g/mol
MDL: MFCD04973773
CAS-Nummer: 351-52-0
UN: 3261
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR