Ethylmagnesiumbromid 3 M in Äther
Lieferant: Thermo Fisher Scientific
Gefahr
87291.JT 87291.JU
87291.JTEA 68.6 EUR
87291.JT 87291.JU
Ethylmagnesiumbromid 3 M in Äther
Ethylmagnesium bromide
CAS No.: 925-90-6 Molecular Formula: C2H5BrMg Formula Weight: 133.28 Physical Form: Liquid Density: 1.020 Sensitivity: Air and Moisture Sensitive Appearance: Pale yellow to brown Odour: Ether-like Melting Point: -116.3° Boiling Point: 34.6° UN No.: 3399 MDL No.: MFCD00000043
Grignard reagent
Formel: C₂H₅BrMg
Molecular Weight: 133.27 g/mol
Siedepunkt: 34.6 °C
Schmelzpunkt: –116.3 °C
Dichte: 1.020
Lagertemperatur: Raumtemperatur
MDL: MFCD00000043
CAS-Nummer: 925-90-6
EINECS: 213-127-3
UN: 3399
ADR: 4.3 I
Bestellen
TECHNISCHE DATEN
Environmentally Preferable