2,6-Dichlor-3-nitrobenzaldehyd

Lieferant: Thermo Scientific

RH01658.250 RH01658.1
MAYBRH01658.250EA 0 EUR
MAYBRH01658.250 MAYBRH01658.1
2,6-Dichlor-3-nitrobenzaldehyd
2,6-Dichloro-3-nitrobenzaldehyde
Formel: C₇H₃Cl₂NO₃
Molecular Weight: 220,0096 g/mol
MDL: MFCD00090332
CAS-Nummer: 5866-97-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR