2-(2-Bromethoxy)benzaldehyd

Lieferant: BIONET RESEARCH

3G-902.1 3G-902.5 3G-902.500
BIOT3G-902.1EA 0 EUR
BIOT3G-902.1 BIOT3G-902.5 BIOT3G-902.500
2-(2-Bromethoxy)benzaldehyd
2-(2-Bromoethoxy)benzaldehyde
Formel: C₉H₉BrO₂
CAS-Nummer: 60633-78-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR