3-(2-Bromethoxy)benzaldehyd

Lieferant: BIONET RESEARCH

Synonyme: 3-(2-Bromoethoxy)benzenecarboxaldehyde

3G-935.500 3G-935.1 3G-935.5 3G-935.10
BIOT3G-935.500EA 0 EUR
BIOT3G-935.500 BIOT3G-935.1 BIOT3G-935.5 BIOT3G-935.10
3-(2-Bromethoxy)benzaldehyd
3-(2-Bromoethoxy)benzaldehyde
Formel: C₉H₉BrO₂
CAS-Nummer: 186191-19-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR