1,4-Dioxan-2,5-dion

Lieferant: Thermo Scientific

Synonyme: Glycolide, 1,4-Dioxane-2,5-dione

SB01616.1 SB01616EA SB01616EE
MAYBSB01616.1EA 0 EUR
MAYBSB01616.1 MAYBSB01616EA MAYBSB01616EE
1,4-Dioxan-2,5-dion
1,4-Dioxan-2,5-dion
Formel: C₄H₄O₄
Molecular Weight: 116,07 g/mol
MDL: MFCD00081108
CAS-Nummer: 502-97-6
EINECS: 207-954-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR