4-Iodbenzaldehyd ≥98%

Lieferant: Thermo Scientific
Achtung

L17448.03 L17448.06 L17448.MD AB180351.0001 AB180351.0005
L17448.03EA 128 EUR
L17448.03 L17448.06 L17448.MD ABCRAB180351.0001 ABCRAB180351.0005
4-Iodbenzaldehyd ≥98%
4-Iodobenzaldehyde

Purity: 98+% CAS No.: 15164-44-0 Molecular Formula: C7H5IO Formula Weight: 232.02 Storage Temperature: 2°C to 8°C Melting Point: 76-78° Beilstein Reference: 636101 MDL No.: MFCD00039576

Formel: C₇H₅IO
Molecular Weight: 232,02 g/mol
Schmelzpunkt: 76…78 °C
MDL: MFCD00039576
CAS-Nummer: 15164-44-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR