2-Brom-5-methoxyphenylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
2-Bromo-5-methoxybenzeneboronic acid, 2-Bromo-5-methoxybenzene boronic acid
BB-2591-5G
BB-2591-1G
COBBBB-2591-5GEA
613
EUR
COBBBB-2591-5G
COBBBB-2591-1G
2-Brom-5-methoxyphenylboronsäure
2-Brom-5-methoxyphenylboronsäure
Formel:
C₇H₈BBrO₃ Molecular Weight: 230.85 g/mol |
MDL:
MFCD06659859 CAS-Nummer: 89694-44-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...