3-Brom-5-methoxyphenylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
(3-Bromo-5-methoxyphenyl)boronic acid
BB-3838-5G
BB-3838-1G
COBBBB-3838-5GEA
353
EUR
COBBBB-3838-5G
COBBBB-3838-1G
3-Brom-5-methoxyphenylboronsäure
3-Brom-5-methoxyphenylboronsäure
Formel:
C₇H₈BBrO₃ Molecular Weight: 230,8516 g/mol |
MDL:
MFCD07369759 CAS-Nummer: 849062-12-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...