5-Brom-2-methoxyphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 5-Bromo-2-methoxybenzeneboronic acid

BB-2605-5G BB-2605-25G BB-2605-1G
COBBBB-2605-5GEA 59.8 EUR
COBBBB-2605-5G COBBBB-2605-25G COBBBB-2605-1G
5-Brom-2-methoxyphenylboronsäure
5-Brom-2-methoxyphenylboronsäure
Formel: C₇H₈BBrO₃
Molecular Weight: 230.85 g/mol
MDL: MFCD00093409
CAS-Nummer: 89694-45-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR