4-Brom-2-methoxyphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (4-Bromo-2-methoxyphenyl)boronic acid

FA-2253-5G FA-2253-1G
COBBFA-2253-5GEA 796 EUR
COBBFA-2253-5G COBBFA-2253-1G
4-Brom-2-methoxyphenylboronsäure
4-Brom-2-methoxyphenylboronsäure
Formel: C₇H₈BBrO₃
Molecular Weight: 230,8516 g/mol
MDL: MFCD11856025
CAS-Nummer: 889849-21-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR