4-Brom-3-methoxyphenylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
(4-Bromo-3-methoxyphenyl)boronic acid
BB-2595-1G
BB-2595-250M
COBBBB-2595-1GEA
703
EUR
COBBBB-2595-1G
COBBBB-2595-250M
4-Brom-3-methoxyphenylboronsäure
4-Brom-3-methoxyphenylboronsäure
Formel:
C₇H₈BBrO₃ Molecular Weight: 230,8500 g/mol |
MDL:
MFCD16036103 CAS-Nummer: 1256345-59-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...