2-Chlor-4-methoxyphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 2-Chloro-4-methoxybenzeneboronic acid

BB-2191-1G BB-2191-5G
COBBBB-2191-1GEA 38.5 EUR
COBBBB-2191-1G COBBBB-2191-5G
2-Chlor-4-methoxyphenylboronsäure
2-Chlor-4-methoxyphenylboronsäure
Formel: C₇H₈BClO₃
Molecular Weight: 186,40 g/mol
MDL: MFCD03411935
CAS-Nummer: 219735-99-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR