2-Chlor-5-methoxyphenylboronsäure ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
2-Chloro-5-methoxyphenylboronic Acid, 2-Chloro-5-methoxybenzeneboronic acid
H53359.03
H53359.06
H53359.03EA
61.8
EUR
H53359.03
H53359.06
2-Chlor-5-methoxyphenylboronsäure ≥98%
2-Chlor-5-methoxyphenylboronsäure
Purity: 98%
CAS No.: 89694-46-2
Molecular Formula: C7H8BClO3
Formula Weight: 186.40
Melting Point: 106-110°
MDL No.: MFCD06659858
Formel:
C₇H₈BClO₃ Molecular Weight: 186,40 g/mol Schmelzpunkt: 106…110 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06659858 CAS-Nummer: 89694-46-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...