2-Chlor-5-methoxyphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 2-Chloro-5-methoxyphenylboronic Acid, 2-Chloro-5-methoxybenzeneboronic acid

BB-2586-5G BB-2586-25G BB-2586-1G
COBBBB-2586-5GEA 154 EUR
COBBBB-2586-5G COBBBB-2586-25G COBBBB-2586-1G
2-Chlor-5-methoxyphenylboronsäure
2-Chlor-5-methoxyphenylboronsäure
Formel: C₇H₈BClO₃
Molecular Weight: 186,40 g/mol
MDL: MFCD06659858
CAS-Nummer: 89694-46-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR