3-Chlor-4-methoxyphenylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
3-Chloro-4-methoxybenzeneboronic acid, (3-Chloro-4-methoxyphenyl)boronic acid
BB-2724-5G
BB-2724-25G
BB-2724-1G
COBBBB-2724-5GEA
153
EUR
COBBBB-2724-5G
COBBBB-2724-25G
COBBBB-2724-1G
3-Chlor-4-methoxyphenylboronsäure
3-Chlor-4-methoxyphenylboronsäure
Formel:
C₇H₈BClO₃ Molecular Weight: 186,40 g/mol |
MDL:
MFCD04039888 CAS-Nummer: 175883-60-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...