5-Chlor-2-methoxyphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (5-Chloro-2-methoxyphenyl)boronic acid, 5-Chloro-2-methoxybenzeneboronic acid

BB-2715-5G BB-2715-25G BB-2715-100G
COBBBB-2715-5GEA 47.1 EUR
COBBBB-2715-5G COBBBB-2715-25G COBBBB-2715-100G
5-Chlor-2-methoxyphenylboronsäure
5-Chlor-2-methoxyphenylboronsäure
Formel: C₇H₈BClO₃
Molecular Weight: 186,40 g/mol
MDL: MFCD01318966
CAS-Nummer: 89694-48-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR