4-Chlor-2-methoxyphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 4-Chloro-2-methoxybenzeneboronic acid, (4-Chloro-2-methoxyphenyl)boronic acid

BB-2197-5G BB-2197-1G
COBBBB-2197-5GEA 324 EUR
COBBBB-2197-5G COBBBB-2197-1G
4-Chlor-2-methoxyphenylboronsäure
4-Chlor-2-methoxyphenylboronsäure
Formel: C₇H₈BClO₃
Molecular Weight: 186,40 g/mol
MDL: MFCD02684317
CAS-Nummer: 762287-57-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR