4-Chlor-3-methoxyphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (4-Chloro-3-methoxyphenyl)boronic acid

BB-2590-5G BB-2590-1G
COBBBB-2590-5GEA 235 EUR
COBBBB-2590-5G COBBBB-2590-1G
4-Chlor-3-methoxyphenylboronsäure
4-Chlor-3-methoxyphenylboronsäure
Formel: C₇H₈BClO₃
Molecular Weight: 186,4000 g/mol
MDL: MFCD01318965
CAS-Nummer: 89694-47-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR