Kaliumtetrachloroaurat ≥98%, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Gold potassium chloride, Potassium tetrachloroaurate(III), Potassium gold(III) chloride

M-Clarity™ Qualitätsstufe = MQ100

334545-1G 334545-5G
SIAL334545-1GEA 189 EUR
SIAL334545-1G SIAL334545-5G
Kaliumtetrachloroaurat ≥98%, Sigma-Aldrich®
Kaliumtetrachloroaurat
Formel: AuCl₄K
Molecular Weight: 377.88 g/mol
MDL: MFCD00043088
CAS-Nummer: 13682-61-6
EINECS: 237-190-1

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR