7-Bromisochinolin

Lieferant: Cayman Chemical

10007081-1 10007081-500
CAYM10007081-1EA 151 EUR
CAYM10007081-1 CAYM10007081-500
7-Bromisochinolin
7-Bromoisoquinoline
Formel: C₉H₆BrN
Molecular Weight: 208,0600 g/mol
MDL: MFCD07368661
CAS-Nummer: 58794-09-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR