Triethylphosphonoacetat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Ethyl 2-(diethoxyphosphoryl)acetate, Diethyl [(ethoxycarbonyl)methyl]phosphonate

M-Clarity™ Qualitätsstufe = MQ100

79525-100ML 79525-500ML
SIAL79525-100MLEA 101 EUR
SIAL79525-100ML SIAL79525-500ML
Triethylphosphonoacetat, Sigma-Aldrich®
Triethylphosphonoacetat
Formel: C₈H₁₇O₅P
Molecular Weight: 224,19 g/mol
MDL: MFCD00009177
CAS-Nummer: 867-13-0
EINECS: 212-757-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR