Triethylphosphonoacetat, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Ethyl 2-(diethoxyphosphoryl)acetate, Diethyl [(ethoxycarbonyl)methyl]phosphonate
M-Clarity™ Qualitätsstufe = MQ200
T61301-100G
T61301-25G
SIALT61301-100GEA
71.1
EUR
SIALT61301-100G
SIALT61301-25G
Triethylphosphonoacetat, Sigma-Aldrich®
Triethylphosphonoacetat
Formel:
C₈H₁₇O₅P Molecular Weight: 224,19 g/mol |
MDL:
MFCD00009177 CAS-Nummer: 867-13-0 EINECS: 212-757-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...