Triethyl-4-phosphonobutyrat ≥97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Triethyl4-phosphonobutyrate
L10488.09
L10488.04
L10488.09EA
117
EUR
L10488.09
L10488.04
Triethyl-4-phosphonobutyrat ≥97%
Triethyl-4-phosphonobutyrat
Purity: 97%
CAS No.: 2327-69-7
Molecular Formula: C10H21O5P
Formula Weight: 252.25
Refractive Index: 1.4370
Boiling Point: 117-119°/0.4mm
MDL No.: MFCD00015168
Formel:
C₁₀H₂₁O₅P Molecular Weight: 252,25 g/mol Siedepunkt: 117…119 °C (0,4 mmHg) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00015168 CAS-Nummer: 2327-69-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...