3-Fluor-L-tyrosin ≥97%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 3-Fluoro-4-hydroxy-L-phenylalanine, (S)-2-Amino-3-(3-fluoro-4-hydroxyphenyl)propanoic acid, H-Tyr(3-F)-OH

L16775.MF L16775.MD
L16775.MFEA 60.1 EUR
L16775.MF L16775.MD
3-Fluor-L-tyrosin ≥97%
3-Fluor-L-tyrosin

Purity: 97% CAS No.: 7423-96-3 Molecular Formula: C9H10FNO3 Formula Weight: 199.18 Melting Point: 260-261° Beilstein Reference: 3204802 UN No.: 2811 MDL No.: MFCD00002607

Formel: C₉H₁₀FNO₃
Molecular Weight: 199,18 g/mol
Schmelzpunkt: 260…261 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00002607
CAS-Nummer: 7423-96-3
UN: 2811
ADR: 6.1,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR