3-Iod-L-tyrosin

Lieferant: BACHEM BIOCHEMICA

Synonyme: H-Tyr(3-I)-OH, (S)-2-Amino-3-(4-hydroxy-3-iodophenyl)propanoic acid

4026261.0005 4026261.0025
BACE4026261.0005EA 0 EUR
BACE4026261.0005 BACE4026261.0025
3-Iod-L-tyrosin
3-Iod-L-tyrosin
Formel: C₉H₁₀INO₃
Molecular Weight: 307,1 g/mol
MDL: MFCD00002608
CAS-Nummer: 70-78-0
EINECS: 200-744-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR