3-Iod-L-tyrosin, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ quality level = MQ200
SIALI8250-1GEA
77.6
EUR
SIALI8250-1G
SIALI8250-25G
SIALI8250-5G
3-Iod-L-tyrosin, Sigma-Aldrich®
3-Iod-L-tyrosin
Formel:
C₉H₁₀INO₃ Molecular Weight: 307,1 g/mol |
MDL:
MFCD00002608 CAS-Nummer: 70-78-0 EINECS: 200-744-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...