1-(3-Bromphenyl)pyrrolidin

Lieferant: COMBI-BLOCKS

HC-6500-5G HC-6500-1G
COBBHC-6500-5GEA 308 EUR
COBBHC-6500-5G COBBHC-6500-1G
1-(3-Bromphenyl)pyrrolidin
1-(3-Bromphenyl)pyrrolidin
Formel: C₁₀H₁₂BrN
Molecular Weight: 226,1130 g/mol
MDL: MFCD04112531
CAS-Nummer: 219928-13-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR