1-(4-Bromphenyl)pyrrolidin

Lieferant: COMBI-BLOCKS

Synonyme: N-(4-Bromophenyl)pyrrolidine

AN-1513-5G AN-1513-1G
COBBAN-1513-5GEA 208 EUR
COBBAN-1513-5G COBBAN-1513-1G
1-(4-Bromphenyl)pyrrolidin
1-(4-Bromphenyl)pyrrolidin
Formel: C₁₀H₁₂BrN
Molecular Weight: 226,1130 g/mol
MDL: MFCD04112480
CAS-Nummer: 22090-26-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR