1-(3-Chlorphenyl)pyrrolidin

Lieferant: COMBI-BLOCKS

HC-6444-1G HC-6444-5G
COBBHC-6444-1GEA 74.2 EUR
COBBHC-6444-1G COBBHC-6444-5G
1-(3-Chlorphenyl)pyrrolidin
1-(3-Chlorphenyl)pyrrolidin
Formel: C₁₀H₁₂ClN
MDL: MFCD09878398
CAS-Nummer: 88154-24-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR