1-(4-Chlorphenyl)-2-pyrrolidinon

Lieferant: FLUOROCHEM

Synonyme: 1-(4-Chlorophenyl)pyrrolidin-2-one

005881-2.5G 005881-10G
FLUO005881-2.5GEA 0 EUR
FLUO005881-2.5G FLUO005881-10G
1-(4-Chlorphenyl)-2-pyrrolidinon
1-(4-Chlorphenyl)-2-pyrrolidinon
Formel: C₁₀H₁₀ClNO
Molecular Weight: 195.6455 g/mol
MDL: MFCD00138486
CAS-Nummer: 7661-33-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR