1-(5-Fluor-2-methylphenyl)pyrrolidin

Lieferant: COMBI-BLOCKS

HC-6443-5G HC-6443-1G
COBBHC-6443-5GEA 149 EUR
COBBHC-6443-5G COBBHC-6443-1G
1-(5-Fluor-2-methylphenyl)pyrrolidin
1-(5-Fluor-2-methylphenyl)pyrrolidin
Formel: C₁₁H₁₄FN
MDL: MFCD09878397
CAS-Nummer: 1000339-32-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR