5-Bromindolin ≥98%

Lieferant: Thermo Scientific
Gefahr

L16992.03 L16992.06
L16992.03EA 44.6 EUR
L16992.03 L16992.06
5-Bromindolin ≥98%
5-Bromindolin

Purity: 98+% CAS No.: 22190-33-6 Molecular Formula: C8H8BrN Formula Weight: 198.06 Melting Point: 36-40° Boiling Point: 114-115°/5mm MDL No.: MFCD00027410

Formel: C₈H₈BrN
Molecular Weight: 198,06 g/mol
Siedepunkt: 114…115 °C (5 mmHg)
Schmelzpunkt: 36…40 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00027410
CAS-Nummer: 22190-33-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR