5-Fluororotsäure Enzymsubstrat

Lieferant: Thermo Fisher Scientific

Synonyme: 5-FOA

R0811 R0812
R0811EA 102 EUR
R0811 R0812
5-Fluororotsäure Enzymsubstrat
5-Fluororotsäure

Thermo Scientific 5-Fluoroorotic Acid (5-fluorouracil-6-carboxylic acid monohydrate; 5-FOA) is used in yeast molecular genetics to detect expression of the URA3 gene, which encodes orotine-5'-monophosphate (OMP) dicarboxylase


Yeast strains carrying a mutation in the URA3 gene grow in the presence of 5-FOA if the media is supplemented with uracil. 5-FOA is frequently used in experiments with Saccharomyces cerevisiae (URA3), Schizosaccharomyces pombe (URA4 and URA5), Candida albicans (URA3), and E. coli (pyrF).


Yeasts with an active URA3 gene (Ura+) convert 5-FOA to fluorodeoxyuridine, which is toxic to cells

Formel: C₅H₃FN₂O₄
Molecular Weight: 174,09 g/mol
MDL: MFCD00042526
CAS-Nummer: 703-95-7
EINECS: 211-876-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR