5-Fluororotsäure ≥98%, blassgelbes kristallines Pulver

Lieferant: MP Biomedicals
Achtung

Synonyme: 5-FOA

0215421050 0215421083 0215421091
ICNA0215421050EA 236 EUR
ICNA0215421050 ICNA0215421083 ICNA0215421091
5-Fluororotsäure ≥98%, blassgelbes kristallines Pulver
5-Fluororotsäure

FOA is a derivative of a pyrimidine precursor and is selectively toxic to yeast cells which synthesize orotidine-5’-phosphate decarboxylase. FOA has been used in a positive selection for uracil-requiring mutants(in the presence of large numbers of wild-type cells) lacking OMPdecase or OPRTase activities. This results in efficient strains for effective transformation systems.


5-Fluoroorotic acid is useful in the selection of orotidine-5′-phosphate decarboxylase mutants of Saccharomyces cerevisiae.It has been used as a selective agent in yeast molecular genetics.It has also been used in the positive selection for uracil auxotrophs of the sulphur-dependent thermophilic archaebacterium sulpholobus acidocaldarius.


Soluble in hot water or methanol; soluble in 1:1 NH4OH : H2O (50 mg/ml with gentle heating).

Formel: C₅H₃FN₂O₄
Molecular Weight: 174,09 g/mol
Schmelzpunkt: 278 °C (Zers., Lit.)
MDL: MFCD00042526
CAS-Nummer: 703-95-7
EINECS: 211-876-0

Bestellen

Spezifikation Testergebnisse

Identity Test Passes
Purity ≥98%
FTIR Conforms to Reference

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR