Ammoniumtetrachloroaurat Hydrat ≥99,99% (Metall-Basis), Premion®
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Ammonium tetrachloroaurate(III) hydrate
40434.03
40434.06
40434.03EA
158
EUR
40434.03
40434.06
Ammoniumtetrachloroaurat Hydrat ≥99,99% (Metall-Basis), Premion®
Ammoniumtetrachloroaurat Hydrat
Purity: 99.99%
CAS No.: 13874-04-9
Molecular Formula: (NH4)AuCl4.xH2O
Formula Weight: 356.82(anhy)
Physical Form: Crystalline powder
Appearance: Yellow-orange
Odour: Odourless
Solubility: Soluble in water and alcohol
UN No.: 3260
MDL No.: MFCD00011503
Formel:
H₄AuCl₄N·H₂O Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011503 CAS-Nummer: 13874-04-9 EINECS: 250-476-0 UN: 3260 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...