Pyrrol-2-carbonsäure, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

P73609-1G P73609-5G
SIALP73609-1GEA 36.6 EUR
SIALP73609-1G SIALP73609-5G
Pyrrol-2-carbonsäure, Sigma-Aldrich®
Pyrrol-2-carbonsäure
Formel: C₅H₅NO₂
MDL: MFCD00005219
CAS-Nummer: 634-97-9
EINECS: 211-221-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR