Ethyl-2-amino-4-phenyl-3-thiophencarboxylat 96%

Lieferant: Thermo Scientific
Achtung

Synonyme: Ethyl 2-Amino-4-phenylthiophene-3-carboxylate

H31867.03 H31867.09
H31867.03EA 23.6 EUR
H31867.03 H31867.09
Ethyl-2-amino-4-phenyl-3-thiophencarboxylat 96%
Ethyl-2-amino-4-phenyl-3-thiophencarboxylat

Purity: 96% CAS No.: 4815-36-5 Molecular Formula: C13H13NO2S Formula Weight: 247.32 Physical Form: Crystalline solid Appearance: Yellow Melting Point: 90-94° MDL No.: MFCD00126391

Formel: C₁₃H₁₃NO₂S
Molecular Weight: 247,32 g/mol
Schmelzpunkt: 90…94 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00126391
CAS-Nummer: 4815-36-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR