Ethyl-2-amino-4-phenyl-3-thiophencarboxylat

Lieferant: Thermo Scientific

Synonyme: Ethyl 2-Amino-4-phenylthiophene-3-carboxylate

HAN00457.1 HAN00457.10
MAYBHAN00457.1EA 0 EUR
MAYBHAN00457.1 MAYBHAN00457.10
Ethyl-2-amino-4-phenyl-3-thiophencarboxylat
Ethyl-2-amino-4-phenyl-3-thiophencarboxylat
Formel: C₁₃H₁₃NO₂S
Molecular Weight: 247,32 g/mol
MDL: MFCD00126391
CAS-Nummer: 4815-36-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR